missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS34 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94087-0.1ml
This item is not returnable.
View return policy
Description
VPS34 Polyclonal antibody specifically detects VPS34 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| VPS34 | |
| Polyclonal | |
| Western Blot 1:100 - 1:500, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200 | |
| EC 2.7.1, EC 2.7.1.137, hVps34, MGC61518, phosphatidylinositol 3-kinase catalytic subunit type 3, Phosphatidylinositol 3-kinase p100 subunit, Phosphoinositide-3-kinase class 3, phosphoinositide-3-kinase, class 3, PI3K type 3, PI3-kinase type 3, PIK3C3, PtdIns-3-kinase type 3, Vps34 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 130-412 of human VPS34 (NP_002638.2). FGKYGMFRQGMHDLKVWPNVEADGSEPTKTPGRTSSTLSEDQMSRLAKLTKAHRQGHMVKVDWLDRLTFREIEMINESEKRSSNFMYLMVEFRCVKCDDKEYGIVYYEKDGDESSPILTSFELVKVPDPQMSMENLVESKHHKLARSLRSGPSDHDLKPNAATRDQLNIIVSYPPTKQLTYEEQDLVWKFRYYLTNQEKALTKFLKCVNWDLPQEAKQALELLGKWKPMDVEDSLELLSSHYTNPTVRRYAVARLRQADDEDLLMYLLQLVQALKYENFDDIK | |
| 0.1 mL | |
| Cancer, Immunology, Innate Immunity, Signal Transduction | |
| 5289 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction