missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VPS33B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551160
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total VPS33B protein.
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene is a member of the Sec-1 domain family, and encodes the human ortholog of rat Vps33b which is homologous to the yeast class C Vps33 protein. The mammalian class C Vps proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway.
Specifications
| VPS33B | |
| Polyclonal | |
| Unconjugated | |
| VPS33B | |
| FLJ14848; hVPS33B; r-vps33b; vacuolar protein sorting 33 homolog B; vacuolar protein sorting 33 homolog B (yeast); vacuolar protein sorting 33B; vacuolar protein sorting 33B (yeast); vacuolar protein sorting 33-like protein B; vacuolar protein sorting homolog r-vps33b; vacuolar protein sorting-associated protein 33B; vacuolar sorting protein 33b; vps33b; VPS33B late endosome and lysosome associated; VPS33B, late endosome and lysosome associated; zgc:110803 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26276 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.3 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9H267 | |
| VPS33B | |
| Fusion protein of human VPS33B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction