missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS29 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-30994
This item is not returnable.
View return policy
Description
VPS29 Polyclonal specifically detects VPS29 in Human samples. It is validated for Western Blot.
Specifications
| VPS29 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 40% Glycerol with 0.01% Thimerosal | |
| VPS29 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human VPS29. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Xenopus laevis (100%) and Xenopus tropicalis (100%). Not yet tested in any other species. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000 | |
| DC15, DC7, DKFZp564F0223, EC 3.1.3.3, FLJ20492, hVPS29, PEP11, PEP11 homolog, retromer protein, vacuolar protein sorting 29 (yeast homolog), vacuolar protein sorting 29 (yeast), vacuolar protein sorting 29 homolog (S. cerevisiae), vacuolar protein sorting-associated protein 29, vacuolar sorting protein VPS29/PEP11, Vesicle protein sorting 29, x 007 protein | |
| Rabbit | |
| 21 kDa | |
| 100 μL | |
| Autophagy, Protein Phosphatase | |
| 51699 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction