missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS28 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38593-20ul
This item is not returnable.
View return policy
Description
VPS28 Polyclonal antibody specifically detects VPS28 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| VPS28 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| ESCRT-I complex subunit VPS28, H-Vps28, MGC60323, vacuolar protein sorting 28 (yeast), vacuolar protein sorting 28 homolog (S. cerevisiae), vacuolar protein sorting-associated protein 28 homolog, yeast class E protein Vps28p homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-221 of human VPS28 (NP_057292.1).,, Sequence:, MFHGIPATPGIGAPGNKPELYEEVKLYKNAREREKYDNMAELFAVVKTMQALEKAYIKDCVSPSEYTAACSRLLVQYKAAFRQVQGSEISSIDEFCRKFRLDCPLAMERIKEDRPITIKDDKGNLNRCIADVVSLFITVMDKLRLEIRAMDEIQPDLRELMETMHRMSHLPPDFEGRQTVSQWLQTLSGMSASDELDDSQVRQMLFDLESAYNAFNRFLHA | |
| 20 μL | |
| Autophagy | |
| 51160 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction