missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS26A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12804-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene belongs to a group of vacuolar protein sorting (VPS) genes. The encoded protein is a component of a large multimeric complex, termed the retromer complex, involved in retrograde transport of proteins from endosomes to the trans-Golgi network. The close structural similarity between the yeast and human proteins that make up this complex suggests a similarity in function. Expression studies in yeast and mammalian cells indicate that this protein interacts directly with VPS35, which serves as the core of the retromer complex. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
| VPS26A | |
| Polyclonal | |
| Unconjugated | |
| VPS26A | |
| HB58, Hbeta58, hVPS26, PEP8A, Vesicle protein sorting 26A, VPS26, VPS26A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 30930, 361846, 9559 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75436, P40336, Q6AY86 | |
| Vps26a | |
| VPS26A Fusion Protein Ag3391 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction