missing translation for 'onlineSavingsMsg'
Learn More

VPS26A Antibody, mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18425636
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425636 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425636 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP275922MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

VPS26A Polyclonal antibody specifically detects VPS26A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS26A
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias FLJ12930, HB58, Hbeta58, hVPS26, PEP8A, vacuolar protein sorting 26 (yeast homolog), vacuolar protein sorting 26 homolog A (S. pombe), vacuolar protein sorting 26 homolog A (yeast), vacuolar protein sorting-associated protein 26A, Vesicle protein sorting 26A, VPS26vacuolar protein sorting 26 (yeast)
Host Species Rabbit
Immunogen Partial synthetic peptide from the C-terminal of human VPS26A (between amino acids 250-320) [UniProt O75436]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy
Primary or Secondary Primary
Gene ID (Entrez) 9559
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.