missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VPS25 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118352
This item is not returnable.
View return policy
Description
VPS25 Polyclonal Antibody for Western Blot, IHC (P), IP
VPS25 (Vacuolar Protein Sorting 25 Homolog) is a Protein Coding gene. Among its related pathways are Delta508-CFTR traffic / ER-to-Golgi in CF and Budding and maturation of HIV virion. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and protein N-terminus binding.
Specifications
| VPS25 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q9BRG1 | |
| VPS25 | |
| E. coli-derived Human VPS25 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| VPS25 | |
| 1110020N13Rik; AW107467; D11Wsu68e; dermal papilla-derived protein 9; DERP9; Eap20; ELL-associated protein of 20 kDa; ESCRT-II complex subunit VPS25; FAP20; HGNC:28122; hVps25; MGC10540; vacuolar protein sorting 25 (yeast); vacuolar protein sorting 25 homolog; vacuolar protein sorting 25 homolog (S. cerevisiae); vacuolar protein-sorting-associated protein 25; VPS25 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 84313 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction