missing translation for 'onlineSavingsMsg'
Learn More

VPS25 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18359212
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18359212 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18359212 Supplier Bio-Techne Supplier No. NBP306483DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

VPS25 Polyclonal antibody specifically detects VPS25 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS25
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Dermal papilla-derived protein 9, DERP9, DERP9hVps25, EAP20, EAP20FAP20, ELL-associated protein of 20 kDa, ESCRT-II complex subunit VPS25, FAP20, MGC10540, vacuolar protein sorting 25 (yeast), vacuolar protein sorting 25 homolog (S. cerevisiae), vacuolar protein-sorting-associated protein 25
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human VPS25 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 84313
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.