missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS24 Rabbit anti-Human, Mouse, Rat, Clone: 1L10B7, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | VPS24 |
|---|---|
| Clone | 1L10B7 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18315583
|
Bio-Techne
NBP3-16902-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18365332
|
Novus Biologicals
NBP3-16902-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VPS24 Monoclonal antibody specifically detects VPS24 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| VPS24 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cell Biology, Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 51652 | |
| IgG | |
| Affinity purified |
| 1L10B7 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CGI-149, CHMP325.1 protein, Chromatin-modifying protein 3, hVps24, member 3,25.1, NEDFFLJ38114, vacuolar protein sorting 24 (yeast), vacuolar protein sorting 24 homolog (S. cerevisiae) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 65-220 of human VPS24 (Q9Y3E7). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title