missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS24 Antibody (SR1492), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22183-100ul
This item is not returnable.
View return policy
Description
VPS24 Monoclonal antibody specifically detects VPS24 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Specifications
| VPS24 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
| SR1492 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
| CGI-149, CHMP325.1 protein, Chromatin-modifying protein 3, hVps24, member 3,25.1, NEDFFLJ38114, vacuolar protein sorting 24 (yeast), vacuolar protein sorting 24 homolog (S. cerevisiae) | |
| A synthesized peptide derived from human VPS24 (Uniprot #: Q9Y3E7) | |
| 100 μg | |
| Autophagy, Cell Biology, Signal Transduction | |
| 51652 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction