missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VPS18 Recombinant Rabbit Monoclonal Antibody (24GB2405)

Rabbit Recombinant Monoclonal Antibody
£126.00 - £434.00
Specifications
| Antigen | VPS18 |
|---|---|
| Clone | 24GB2405 |
| Concentration | 0.31 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30276024
![]() |
Invitrogen™
MA557319 |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275306
|
Invitrogen™
MA557320 |
20 μL |
£126.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VPS18 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
VPS18 is a vesicle mediated protein that plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human homolog of yeast class C Vps18 protein. The mammalian class C Vps proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway.Specifications
| VPS18 | |
| 0.31 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8R307, Q9P253 | |
| 228545, 296083, 57617 | |
| A synthesized peptide derived from human VPS18 (aa 1-28). | |
| Antibody |
| 24GB2405 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| VPS18 | |
| 9930024E13Rik; hVPS18; KIAA1475; PEP3; vacuolar protein sorting 18 (yeast); vacuolar protein sorting 18 homolog; vacuolar protein sorting 18 homolog (S. cerevisiae); vacuolar protein sorting protein 18; vacuolar protein sorting-associated protein 18 homolog; Vps18; VPS18 CORVET/HOPS core subunit; VPS18, CORVET/HOPS core subunit | |
| VPS18 | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title