missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS18 Rabbit anti-Human, Mouse, Rat, Clone: 1W8T9, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | VPS18 |
|---|---|
| Clone | 1W8T9 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18302795
|
Novus Biologicals
NBP3-15542-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18320885
|
Novus Biologicals
NBP3-15542-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VPS18 Monoclonal antibody specifically detects VPS18 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| VPS18 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cellular Markers | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 57617 | |
| IgG | |
| Affinity purified |
| 1W8T9 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| hVPS18, KIAA1475PEP3, vacuolar protein sorting 18 homolog (S. cerevisiae), vacuolar protein sorting protein 18, vacuolar protein sorting-associated protein 18 homolog | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human VPS18 (Q9P253). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title