missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS16, Mouse anti-Human, Clone: 2F10, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00064601-M05.100ug
This item is not returnable.
View return policy
Description
Sequence: IGYLPFVEICMKQHNKYEAKKYASRVGPEQKVKALLLVGDVAQAADVAIEHRNEAELSLVLSHCTGATDGATADKIQRARAQAQKKSpecifications
| VPS16 | |
| Monoclonal | |
| Unconjugated | |
| vacuolar protein sorting 16 homolog (S. cerevisiae) | |
| hVPS16 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 2F10 | |
| In 1x PBS, pH 7.4 | |
| NM_022575 | |
| VPS16 | |
| VPS16 (NP_072097.2, 754 a.a. ∼ 839 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 64601 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction