missing translation for 'onlineSavingsMsg'
Learn More

VPS13B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£294.00 - £460.00

Specifications

Antigen VPS13B
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18238384
View Documents
Novus Biologicals
NBP2-55104
100 μL
£460.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18600477
View Documents
Novus Biologicals
NBP2-55104-25ul
25 μL
£294.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

VPS13B Polyclonal specifically detects VPS13B in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

VPS13B
Polyclonal
Rabbit
Human
cohen syndrome protein 1, DKFZp313I0811, KIAA0532, vacuolar protein sorting 13 homolog B (yeast)
VPS13B
IgG
Affinity Purified
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
157680
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PSVIKIHTLVESLKLSITDQQLPMFIRIMQLGIALYYGEIGNFKEGEIEDLTCHNKDMLGNITGSEDETR
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Certificates

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.