missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
VPS11 Polyclonal antibody specifically detects VPS11 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | VPS11 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 405 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | hVPS11, PEP5, RING finger protein 108, RNF108END1, vacuolar protein sorting 11 (yeast homolog), vacuolar protein sorting 11 homolog (S. cerevisiae), vacuolar protein sorting protein 11, vacuolar protein sorting-associated protein 11 homolog |
| Host Species | Rabbit |
| Immunogen | The immunogen recognized by this antibody maps to a region between residue 891 and 941 of human Vacuolar Protein Sorting 11 using the numbering given in entry NP_068375.3 (GeneID 55823). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?