missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
VPREB3 Monoclonal antibody specifically detects VPREB3 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | VPREB3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI1A3 |
| Conjugate | DyLight 550 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | N27C7-2, pre-B lymphocyte 3,8HS20, pre-B lymphocyte gene 3, pre-B lymphocyte protein 3, Protein VPreB3 |
| Host Species | Mouse |
| Immunogen | Human recombinant protein fragment corresponding to amino acids 48-123 of human VPREB3 (NP_037510) produced in E.coli. |
| Purification Method | >95% |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?