missing translation for 'onlineSavingsMsg'
Learn More

VPREB3 Antibody (OTI1A3) [Alexa Fluor™ 700], Novus Biologicals™

Product Code. 30688666 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30688666 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688666 Supplier Novus Biologicals Supplier No. NBP345027AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

VPREB3 Monoclonal antibody specifically detects VPREB3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPREB3
Applications Western Blot
Classification Monoclonal
Clone OTI1A3
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias N27C7-2, pre-B lymphocyte 3,8HS20, pre-B lymphocyte gene 3, pre-B lymphocyte protein 3, Protein VPreB3
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 48-123 of human VPREB3 (NP_037510) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 29802
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.