missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPREB1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-49200
This item is not returnable.
View return policy
Description
VPREB1 Polyclonal antibody specifically detects VPREB1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| VPREB1 | |
| Polyclonal | |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| CD179 antigen-like family member A, CD179a, IGI, immunoglobulin iota chain, pre-B lymphocyte 1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: QPVLHQPPAMSSALGTTIRLTCTLRNDHDIGVYSVYWYQQRPGHPPRFLLRYFSQSDKSQGPQVPPRFSGSKDVARNRGYLSISELQPEDEAMYYCAMGARSSEKEEREREWEEEMEPT | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 7441 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido