missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VprBP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£342.00 - £470.00
Specifications
| Antigen | VprBP |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18483042
|
Novus Biologicals
NBP1-85284-25ul |
25ul |
£342.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18236268
|
Novus Biologicals
NBP1-85284 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
VprBP Polyclonal specifically detects VprBP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Tekniske data
| VprBP | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 9730 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:FTCCAFSARERFLMLGTCTGQLKLYNVFSGQEEASYNCHNSAITHLEPSRDGSLLLTSATWSQPLSALWGMKSVFDMKHSFTEDHYVEFSKHSQDRVIG | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| DCAF1DDB1 and CUL4 associated factor 1, DDB1- and CUL4-associated factor 1, HIV-1 Vpr-binding protein, KIAA0800protein VPRBP, MGC102804, RIP, Vpr (HIV-1) binding protein, Vpr-binding protein, VprBP, Vpr-interacting protein | |
| VPRBP | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel