Learn More
Invitrogen™ VPARP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120485
Description
Positive test controls include: HeLa, Mouse heart. The target is usually found in the following locations: Cytoplasm, Nucleus, cytoskeleton, spindle. Immunogen sequence: MVMGIFANCI FCLKVKYLPQ QQKKKLQTDI KENGGKFSFS LNPQCTHIIL DNADVLSQYQ LNSIQKNHVH IANPDFIWKS IREKRLLDVK NYDPYKPLDI TPPPDQKASS SEVKTEGLCP DSATEEEDTV ELTEFGMQNV EIPHLPQDFE VAKYNTLEKV GMEGGQEAVV VELQCSRDSR DCPFLISSHF LLDDGMETRR QFAIKKTSED ASEYFENYIE ELKKQGFLLR EHFTPEATQL ASEQLQALLL EEVMNSSTLS QEVSDLVEMI WAEALGHLEH MLLKPVNRIS LNDVSKAEGI.
This gene encodes poly(ADP-ribosyl)transferase-like 1 protein, which is capable of catalyzing a poly(ADP-ribosyl)ation reaction. This protein has a catalytic domain which is homologous to that of poly (ADP-ribosyl) transferase, but lacks an N-terminal DNA binding domain which activates the C-terminal catalytic domain of poly (ADP-ribosyl) transferase. Since this protein is not capable of binding DNA directly, its transferase activity may be activated by other factors such as protein-protein interaction mediated by the extensive carboxyl terminus.
Specifications
| VPARP | |
| Polyclonal | |
| Unconjugated | |
| PARP4 | |
| 193 kDa vault protein; ADP-ribosyltransferase (NAD+; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 1; ADP-ribosyltransferase diphtheria toxin-like 4; ADPRTL1; ARTD4; C030027K23Rik; E230037B21Rik; Gm743; H5 proline-rich; I-alpha-I-related; KIAA0177; p193; Parp4; PARP-4; PARPL; PARP-related; PARP-related/IalphaI-related H5/proline-rich; PH5P; poly (ADP-ribose) polymerase family, member 4; poly (ADP-ribose) polymerase)-like 1; poly [ADP-ribose] polymerase 4; poly(ADP-ribose) polymerase family member 4; poly(ADP-ribose) synthetase; poly(ADP-ribosyl)transferase-like 1; Protein mono-ADP-ribosyltransferase PARP4; Vault poly(ADP-ribose) polymerase; vault protein, 193-kDa; VAULT3; von Willebrand factor A domain containing 5C; VPARP; VWA5C | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 143, 328417, 361046 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.75 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UKK3 | |
| PARP4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human PARP4 (NP_006428.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.