missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ VP-SFM AGT™ Medium

Product Code. p-7076324
Click to view available options
Quantity:
1 L
10 kg
10 L
100 L
Unit Size:
1 L
10 L
100 L
10kg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10514043

Brand: Gibco™ 12559027

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Gibco VP-SFM AGT Medium is a flexible-format medium that is equivalent to VP-SFM with the addition of Advanced Granulation Technology for simplified preparation.

Gibco VP-SFM AGT Medium is a flexible-format medium that is equivalent to VP-SFM with the addition of Advanced Granulation Technology for simplified preparation. VP-SFM is a serum-free, animal-origin-free culture medium designed for growth of VERO cells and virus production. It is also suitable for the growth of COS-7, MDCK, BHK-21, and Hep2 cells and for recombinant protein production.

VP-SFM AGT™ is a serum-free, ultra-low protein medium in an easy-to-use granular format. It is designed for the growth of VERO cells for virus production and for the growth of COS-7, MDCK, BHK-21 and Hep-2 cells for recombinant protein production.
Advanced Granulation Technology = AGT
TRUSTED_SUSTAINABILITY

Specifications

Cell Line MDCK, VERO, HEp2, BHK-21, COS-7
Classification Animal Origin-free, Low Protein, Serum-free
Form Powder (AGT)
Product Type Serum Free Medium (SFM)
Serum Level Serum-free
With Additives Phenol Red, Sodium Pyruvate, Sodium Bicarbonate
Without Additives No Glutamine
Product Line Gibco
Quantity 1 L
Shipping Condition Wet Ice
Endotoxin Level Low
Format Securitainer Capped Vial
Included Antibiotics No antibiotics
Reconstituted Volume 1 L
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Adherent Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.