missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vortioxetine Lactose Adduct, TRC
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
Specifications
Specifications
| Chemical Name or Material | Vortioxetine Lactose Adduct |
| Analyte or Component Names | Vortioxetine Lactose Adduct |
| Shipping Condition | Room Temperature |
| Molecular Formula | C30H42N2O10S |
| Synonym | N-[(3S)-1-[(Dimethylamino)carbonyl]-3-pyrrolidinyl]-5-[(Z)-(5-fluoro-1,2-dihydro-2-oxo-3H-indol-3-ylidene)methyl]-2,4-dimethyl-1H-pyrrole-3-carboxamide |
| InChI Formula | InChI=1S/C30H42N2O10S/c1-16-7-8-21(17(2)13-16)43-22-6-4-3-5-18(22)31-9-11-32(12-10-31)29-26(38)25(37)28(20(15-34)40-29)42-30-27(39)24(36)23(35)19(14-33)41-30/h3-8,13,19-20,23-30,33-39H,9-12,14-15H2,1-2H3 |
| Solution Type | Neat |
| SMILES | OC(C(O)C(OC1OC(CO)C(O)C(O)C1O)C(CO)O2)C2N3CCN(CC3)C4=C(C=CC=C4)SC(C=CC(C)=C5)=C5C |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 622.73 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction