missing translation for 'onlineSavingsMsg'
Learn More

Vortioxetine.HBr, 1mg/ml in Methanol (as free base), Lipomed

Product Code. 30458859
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458859 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458859 Supplier Lipomed Supplier No. LPMVOX1884HB1LM

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Vortioxetine.HBr, 1mg/ml in Methanol (as free base)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vortioxetine Hydrobromide
CAS 960203-27-4
Analyte or Component Names Vortioxetine Hydrobromide
Shipping Condition Room Temperature
Molecular Formula C18 H22 N2 S . Br H
Concentration 1 mg/mL
Synonym Piperazine, 1-[2-[(2,4-dimethylphenyl)thio]phenyl]-, hydrobromide (1:1), 1-[2-[(2,4-Dimethylphenyl)thio]phenyl]piperazine hydrobromide, 1-[2-(2,4-Dimethylphenylsulfanyl)phenyl]piperazine hydrobromide, Lu AA 21004 hydrobromide, Vortioxetine hydrobromide
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Br.Cc1ccc(Sc2ccccc2N3CCNCC3)c(C)c1
IUPAC Name 1-[2-(2,4-dimethylphenyl)sulfanylphenyl]piperazine;hydrobromide
Solvent or Matrix Methanol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 379.358
Quantity 1 mL
Formula Weight 378.077 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.