missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ Vortexer Mixer, Blue

The Vortexer is a multi-purpose, and maximum convenience unit. This sleek and modern designed vortex mixer is capable of meeting the mixing requirements of the most popular test tubes used in today’s lab without the need to swap out mixing head units.

Brand:  Heathrow Scientific™ HS120212

238.04 GBP valid until 2024-04-30
Use promo code "21656" to get your promotional price.



Code : NEW

Additional Details : Weight : 4.85000kg

 View more versions of this product

Product Code. 15599811

  • £255.00 / Each
Estimated Shipment: 09-08-2024
to see stock.
Add to basket

Description

Description

The innovative design makes maximum use of the space available, providing holding slots for no less than six different sizes of tube and a standard assay plate, while the center pad area can be used for quick hand-held mixing of a vast range of tubes and vessels.

  • Body made from durable ABS to resist most commonly used chemicals, and is easy to clean
  • Stable elastomeric device feet, provide vibration dampening and prevent movement during operation
  • Innovative head can be removed for cleaning purposes
  • US Design Patent No. D787,085, US Utility Patent No. 9895670

Specifications

Specifications

3.7 mm
1000 to 3000 rpm
220/240 V
0.4 A
Vortexing unit
50/60 Hz
7.41 in.
6.7 x 7.5 in.
19.7 cm
5.51 in.
140 mm
4.5 in.
11.43 cm
5 kg
3.93 kg
HS120212
1 x AC power cord, 1 x mixing head, 3 x elastic retention cords, 3 x central pads
3-Modes: 1000, 2000, and 3000 rpm
135 g
220/240 V, 50/60 Hz
44 W
50 mL, 15 mL, 1.5/2 mL, 0.5 mL, and 0.2 mL
CE
ABS Base, Nylon Platform
19 cm
7.68 in.
6.7 in.
17.1 cm
6.67 in.
17.1 cm
11.12 lb.
8.66 lb.
Vortexer Mixer 220/240 V, CE Plug
Tubes, Assay Plates
Brushless
5 Years
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers