Learn More
Heathrow Scientific™ Vortexer Mixer, Blue
The Vortexer is a multi-purpose, and maximum convenience unit. This sleek and modern designed vortex mixer is capable of meeting the mixing requirements of the most popular test tubes used in today’s lab without the need to swap out mixing head units.
Brand: Heathrow Scientific™ HS120214
276.73 GBP valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Code : 46
Additional Details : Weight : 4.85000kg
Description
The innovative design makes maximum use of the space available, providing holding slots for no less than six different sizes of tube and a standard assay plate, while the center pad area can be used for quick hand-held mixing of a vast range of tubes and vessels.- Body made from durable ABS to resist most commonly used chemicals, and is easy to clean
- Stable elastomeric device feet, provide vibration dampening and prevent movement during operation
- Innovative head can be removed for cleaning purposes
- US Design Patent No. D787,085, US Utility Patent No. 9895670
Specifications
3.7 mm | |
1000 to 3000 rpm | |
220/240 V | |
0.4 A | |
Vortexing unit | |
UK Plug | |
ABS Base, Nylon Platform | |
19 cm | |
6.7 x 7.5 x 7.8 in. | |
19.7 cm | |
5.51 in. | |
140 mm | |
4.5 in. | |
11.43 cm | |
5 kg | |
3.93 kg | |
Vortexer Mixer 220/240 V, UK Plug | |
Tubes, Assay Plates | |
Brushless | |
5 Years |
135 g | |
220/240 V, 50/60 Hz | |
44 W | |
50 mL, 15 mL, 1.5/2 mL, 0.5 mL, and 0.2 mL | |
SGS-US-C, CE, CB Scheme, RoHS 2, PSE,and WEEE | |
50/60 Hz | |
7.41 in. | |
6.7 x 7.5 in. | |
7.68 in. | |
6.7 in. | |
17.1 cm | |
6.67 in. | |
17.1 cm | |
11.12 lb. | |
8.66 lb. | |
3 position slider (top) intermittent, (middle) off, (bottom) continuous | |
HS120214 | |
1 x AC power cord, 1 x mixing head, 3 x elastic retention cords, 3 x central pads | |
3-Modes: 1000, 2000, and 3000 rpm |