Learn More
Corning™ LSE™ Digital Vortexer Platform
Product Code. 15975911 Shop All Corning Products
Change view
Click to view available options
Product Type:
Platform
Platform with non-stick mat
Spring Loaded Platform
Unit Size:
Each
Product Code. Product Type unitSize
15975911 Platform with non-stick mat Each
15965911 Platform Each
15985911 Spring Loaded Platform Each
3 options
Product Code. 15975911
Brand: Corning™ 48011
Only null left
Add to basket
Add to basket
Corning™ Vortexer Platform for use with Corning™ LSE Multiposition Digital Vortexer.
The Corning Digital Vortexer is a multipurpose shaker dedicated to a variety of shaking and mixing applications. Three easily removable and interchangeable platforms accommodate many different sample containers such as flasks, bottles, tube racks, plates, boxes, and dishes. The clamp platform has spring loaded retaining bars that adjust to fit vessels of different shapes and sizes. The microplate platform is designed to securely hold up to four microplates. Operation of the vortexer can be timed or continuous, while the speed is variable. The broad speed range allows the vortexer to be used for gentle mixing as well as vigorous agitation. A powerful motor, and a chemically resistant and easy to clean surface makes the equipment a perfect choice for years of trouble-free use.
Features and Benefits
  • Three versatile platform options (available separately)
  • Compact design with easy-access control panel
  • Non-slip rubber mat to keep containers in place
  • Safe for cold room or incubator use
  • Five-year warranty
TRUSTED_SUSTAINABILITY
Specifications
Product Type Platform with non-stick mat
Description Non-slip rubber mat to keep containers in place
For Use With (Application) General Purpose Mixing
For Use With (Equipment) LSE Multiposition Digital Vortexer