Learn More
Cole-Parmer™ Vortex-Genie™ 2 Shaker
Adapt your shaker to a variety of applications. Cole-Parmer™ Vortex-Genie™ 2 Shaker features variable speed shaking which allows you to adjust the level of mixing from gentle shaking to vigorous vortexing.
Brand: Cole-Parmer™ 04724-05
Code : A0
Additional Details : Weight : 0.50000kg
Description
Shaker allows you to control speed intensity by varying pressure to the head. Use the pop-off cup for single tube shaking or the 3 in. platform for multiple tube or flask shaking. Enhanced vortex action for maximum high-speed touch mixing. Solid metal casing with rubber feet prevents shaker from ''walking'' across your benchtop. Includes pop-off cup and 3 in. platform. Power cord and plug (Euro plug on 230 VAC model).Specifications
600 to 2700 rpm | |
220 V | |
50 Hz | |
123.8 mm |
220 V, 50 Hz | |
Shaker | |
152.4 mm |