missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VorTemp™ 56 Shaking Incubator and Temperature Validation Kit
Description
The VorTemp 56 microplate shaking incubator is designed for simultaneous heating and mixing of QuantiGene Plex assays. The temperature is adjustable over a broad range, and mechanical convection provides a uniform and stable environment within the chamber.
VorTemp features:
- A microprocessor to control all parameters including temperature and timed operation
- Self-diagnostic software to continuously monitor for proper temperature and shaking speed, alerting the user to any errors
- Heating and shaking that are controlled by feedback loops which precisely maintain chosen parameters, independent of load or voltage fluctuations
- Sturdy construction for stability during high speed shaking
- An insulated chamber to maintain temperature and reduce operating noise
Specifications
Specifications
| Voltage | 120 V |
| Temperature Range (Metric) | 4°C to 99.5°C |
| Product Type | Shaking Incubator |
| Material | Coated Steel |
| Humidity Range | 85% |
| Includes | 1 VorTemp 56 Shaking Incubator, 1 QuantiGene Temperature Validation Kit |
| Quantity | 1 Ea. |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?