missing translation for 'onlineSavingsMsg'
Learn More

Voreloxin Hydrochloride, MedChemExpress

Product Code. 16497068
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497068 10 mg 10mg
16449617 5 mg 5mg
16423117 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16497068 Supplier Medchem Express Supplier No. HY16518

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Voreloxin Hydrochloride is a first-in-class topoisomerase II inhibitor that intercalates DNA and induces site-selective DNA DSB, G2 arrest, and apoptosis.

Voreloxin Hydrochloride is a first-in-class topoisomerase II inhibitor that intercalates DNA and induces site-selective DNA DSB, G2 arrest, and apoptosis.

Specifications

Chemical Name or Material Voreloxin Hydrochloride
CAS 175519-16-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C18H20ClN5O4S
Quantity 10 mg
Synonym SNS-595 Hydrochloride Vosaroxin Hydrochloride AG 7352 Hydrochloride
Solubility Information DMSO : 2 mg/mL (4.57 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 2 mg/mL (4.57 mM; ultrasonic and warming and heat to 60°C)
SMILES [H]Cl.O=C(C1=CN(C2=NC=CS2)C3=C(C=CC(N4C[C@H](OC)[C@@H](NC)C4)=N3)C1=O)O
Molecular Weight (g/mol) 437.9
Formula Weight 437.9
Percent Purity 99.29%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.