missing translation for 'onlineSavingsMsg'
Learn More

Vonoprazan, MedChemExpress

Product Code. 16454196
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16454196 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16454196 Supplier Medchem Express Supplier No. HY100007

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vonoprazan (TAK-438 free base), a proton pump inhibitor (PPI), is a potent and orally active potassium-competitive acid blocker (P-CAB), with antisecretory activity.

Vonoprazan inhibits H+,K+-ATPase activity in porcine gastric microsomes with an IC50 of 19 nM at pH 6.5. Vonoprazan is developed for the research of acid-related diseases, such as gastroesophageal reflux disease and peptic ulcer disease. Vonoprazan can be used for eradication of Helicobacter pylori.

Specifications

Chemical Name or Material Vonoprazan
CAS 881681-00-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C17H16FN3O2S
Quantity 100 mg
Synonym TAK-438 free base
Solubility Information DMSO : 100 mg/mL (289.53 mM; Need ultrasonic)
SMILES O=S(N1C=C(CNC)C=C1C2=CC=CC=C2F)(C3=CC=CN=C3)=O
Molecular Weight (g/mol) 345.39
Formula Weight 345.39
Percent Purity 99.61%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.