missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vonoprazan Fumarate, TRC
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Specifications
Specifications
| Chemical Name or Material | Vonoprazan Fumarate |
| CAS | 881681-01-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C17 H16 F N3 O2 S . C4 H4 O4 |
| Quantity | 50 mg |
| Synonym | 1H-Pyrrole-3-methanamine','5-(2-fluorophenyl)-N-methyl-1-(3-pyridinylsulfonyl)-','(2E)-2-butenedioate (1:1), 5-(2-Fluorophenyl)-N-methyl-1-(3-pyridinylsulfonyl)-1H-pyrrole-3-methanamine (2E)-2-butenedioate (1:1), TAK 438, TAK 438 monofumarate, Takecab, Vocinti, Vonoprazan fumarate, 5-(2-Fluorophenyl)-N-methyl-1-(3-pyridinylsulfonyl)-1H-pyrrole-3-methanamine fumarate, 1-[5-(2-Fluorophenyl)-1-[(pyridin-3-yl)sulfonyl]-1H-pyrrol-3-yl]-N-methylmethanamine fumarate, 1H-Pyrrole-3-methanamine','5-(2-fluorophenyl)-N-methyl-1-(3-pyridinylsulfonyl)-','(2E)-2-butenedioate (1:1), 5-(2-Fluorophenyl)-N-methyl-1-(3-pyridinylsulfonyl)-1H-pyrrole-3-methanamine (2E)-2-butenedioate (1:1), TAK 438, TAK 438 monofumarate, Takecab, Voci |
| InChI Formula | InChI=1S/C17H16FN3O2S.C4H4O4/c1-19-10-13-9-17(15-6-2-3-7-16(15)18)21(12-13)24(22,23)14-5-4-8-20-11-14;5-3(6)1-2-4(7)8/h2-9,11-12,19H,10H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1+ |
| SMILES | CNCc1cc(c2ccccc2F)n(c1)S(=O)(=O)c3cccnc3.OC(=O)\C=C\C(=O)O |
| IUPAC Name | (E)-but-2-enedioic acid;1-[5-(2-fluorophenyl)-1-pyridin-3-ylsulfonylpyrrol-3-yl]-N-methylmethanamine |
| Molecular Weight (g/mol) | 461.463 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction