missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vonoprazan Fumarate, MedChemExpress
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Vonoprazan Fumarate (TAK-438), a proton pump inhibitor (PPI), is a potent and orally active potassium-competitive acid blocker (P-CAB), with antisecretory activity. Vonoprazan Fumarate inhibits H+,K+-ATPase activity in porcine gastric microsomes with an IC50 of 19 nM at pH 6.5. Vonoprazan Fumarate is developed for the research of acid-related diseases, such as gastroesophageal reflux disease and peptic ulcer disease.
Specifications
Specifications
| Chemical Name or Material | Vonoprazan Fumarate |
| CAS | 881681-01-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H20FN3O6S |
| Quantity | 100 mg |
| Synonym | TAK-438 |
| Solubility Information | DMSO : 50 mg/mL (108.35 mM; Need ultrasonic) |
| SMILES | O=C(O)/C=C/C(O)=O.O=S(N1C=C(CNC)C=C1C2=CC=CC=C2F)(C3=CC=CN=C3)=O.[(E)] |
| Molecular Weight (g/mol) | 461.46 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction