missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Vonlerizumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA559896
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Vonlerizumab, also known as an anti-human CD134 (OX40) monoclonal antibody, is extensively studied for its targeted immunomodulatory function. TNFRSF4, also known as OX40 or CD134, is a gene encoding a protein that belongs to the tumor necrosis factor receptor (TNFR) superfamily. This receptor is primarily expressed on activated CD4 and CD8 T cells and serves as a co-stimulatory molecule involved in the regulation of immune responses. Upon binding with its ligand, OX40L (CD252), OX40 transmits signals that promote T cell proliferation, survival, and cytokine production, thereby enhancing the immune response. The expression of the OX40 gene is tightly controlled and can be upregulated by chromatin remodeling in T cells. OX40 signaling plays a crucial role in the accumulation of protective T cells during anti-viral and anti-tumor immune responses and is being explored as a therapeutic target in both autoimmune diseases and cancer immunotherapy due to its ability to modulate T cell-mediated immune functions.
Specifications
| Vonlerizumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction