missing translation for 'onlineSavingsMsg'
Learn More
Learn More
von Willebrand Factor Mouse, Clone: F8/86, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 7450-MSM6-P1
This item is not returnable.
View return policy
Specifications
| von Willebrand Factor | |
| Cytoplasmic | |
| F8/86 | |
| Unconjugated | |
| NP_000543 | |
| Mouse | |
| 250kDa | |
| Protein A/G Purified | |
| RUO | |
| 7450 | |
| IgG1 |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| VWF | |
| von Willebrand Factor isolated from human plasma | |
| HUVEC cells or Tonsil | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction