missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Von Willebrand Factor Mouse anti-Human, Clone: rVWF/2480, Novus Biologicals™
Mouse Monoclonal Antibody
£197.00
Specifications
| Antigen | Von Willebrand Factor |
|---|---|
| Clone | rVWF/2480 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
Description
Von Willebrand Factor Monoclonal specifically detects Von Willebrand Factor in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| Von Willebrand Factor | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF | |
| Recombinant full-length human Von Willebrand Factor protein | |
| Primary | |
| Store at 4°C. |
| rVWF/2480 | |
| Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 7450 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title