missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Von Willebrand Factor Antibody (VWF/7979R) [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-21110JF669
This item is not returnable.
View return policy
Description
Von Willebrand Factor Monoclonal antibody specifically detects Von Willebrand Factor in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Von Willebrand Factor | |
| Monoclonal | |
| Janelia Fluor 669 | |
| coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF | |
| Recombinant fragment (around aa 1150-1250) of human Von Willebrand Factor protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer | |
| 7450 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| VWF/7979R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction