missing translation for 'onlineSavingsMsg'
Learn More

Von Willebrand Factor Antibody (VWF/1465), DyLight 405, Novus Biologicals™

Product Code. 18429763 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429763 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429763 Supplier Novus Biologicals Supplier No. NBP254430V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Von Willebrand Factor Monoclonal antibody specifically detects Von Willebrand Factor in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Von Willebrand Factor
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone VWF/1465
Conjugate DyLight 405
Formulation 50 mM Sodium Borate
Gene Alias coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF
Host Species Mouse
Immunogen Recombinant fragment of human Von Willebrand Factor protein (aa1815-1939) (exact sequence is proprietary) (Uniprot: P04275)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7450
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.