missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Von Willebrand Factor Monoclonal specifically detects Von Willebrand Factor in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | Von Willebrand Factor |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | CL1950 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:1000, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Gene Accession No. | P04275 |
| Gene Alias | coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF |
| Gene Symbols | VWF |
| Host Species | Mouse |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?