missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Von Hippel Lindau Antibody (AT82B10), mFluor Violet 500 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Von Hippel Lindau Monoclonal antibody specifically detects Von Hippel Lindau in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Applications | Western Blot, ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Conjugate | mFluor Violet 500 SE |
| Host Species | Mouse |
| Regulatory Status | RUO |
| Gene ID (Entrez) | 7428 |
| Content And Storage | Store at 4C in the dark. |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?