missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Von Hippel Lindau Antibody (AT82B10), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04355C
This item is not returnable.
View return policy
Description
Von Hippel Lindau Monoclonal specifically detects Von Hippel Lindau in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Von Hippel Lindau | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| VHL | |
| Recombinant human His-VHL (1-154 aa) purified from E. coli | |
| 0.1 mL | |
| Apoptosis, Cancer, HIF Target Genes, Hypoxia, Neuronal Cell Markers, Neuronal Stem Cell Markers | |
| 7428 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| AT82B10 | |
| Western Blot, ELISA | |
| elongin binding protein, HRCA1, Protein G7, pVHL, RCA1, VHL1, von Hippel-Lindau disease tumor suppressor, von Hippel-Lindau syndrome, von Hippel-Lindau tumor suppressor | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human VHL mAb, clone 52A11, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human His-VHL protein. | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction