missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Von Hippel Lindau Antibody (AT82B10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04355
This item is not returnable.
View return policy
Description
Von Hippel Lindau Monoclonal specifically detects Von Hippel Lindau in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Von Hippel Lindau | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:1000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence | |
| elongin binding protein, HRCA1, Protein G7, pVHL, RCA1, VHL1, von Hippel-Lindau disease tumor suppressor, von Hippel-Lindau syndrome, von Hippel-Lindau tumor suppressor | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Anti-human VHL mAb, clone 52A11, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human His-VHL protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| AT82B10 | |
| Unconjugated | |
| NP_000542 | |
| VHL | |
| Recombinant human His-VHL (1-154 aa) purified from E. coli | |
| 0.1 mL | |
| Apoptosis, Cancer, HIF Target Genes, Hypoxia, Neuronal Cell Markers, Neuronal Stem Cell Markers | |
| 7428 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction