Learn More
Kartell SpA Volumetric Flask with Cap
Product Code. 10565811
Change view
Click to view available options
Capacity (Metric):
50 mL
250 mL
500 mL
1000 mL
Unit Size:
Pack of 10
Pack of 20
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10565811 250 mL Pack of 10
10458652 50 mL Pack of 20
10051071 500 mL Pack of 10
10011111 1000 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10565811 Supplier Kartell SpA Supplier No. 0248700
Only null left
Add to basket
Add to basket
Volumetric flask with cap, PMP
Crystal clear volumetric flask creates no meniscus so final readings are easy. Calibrated gravimetrically at 20°C. Thick walls eliminate distortion. Thin necks make final measurement precise. Autoclavable to 170°C. Chemically non-absorbant. No wetting. Stopper included
Specifications
Capacity (Metric) 250 mL
Material PMP
Product Type Volumetric Flask
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Mouth Style Threaded
Sterility Non-sterile
Bottom Shape Round
Surface Treatment Untreated