Learn More
KARTELL Volumetric Flask
Product Code. 10681144
Change view
Click to view available options
Capacity (Metric):
50 mL
250 mL
500 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10681144 50 mL Each
10335063 250 mL Each
10040023 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10681144 Supplier KARTELL Supplier No. 0169600
Volumetric Flask, made of transparent fluorinated resin, has high chemical and temperature resistance.
  • Volumetric flask made of transparent fluorinated resin with high chemical and temperature resistance (-250°C to +270°C).
  • Supplied with standard DIN stoppers in PFA.
  • Individually calibrated ringmark according to DIN EN ISO 1042, class “A”, Standards.
  • Laser engraved lot number.
Specifications
Capacity (Metric) 50 mL
Material PFA
Autoclavable Not Autoclavable
Disposable Reusable
Graduated No
Mouth Style Narrow
Sterility Non-sterile
Bottom Shape Flat
Surface Treatment Non-treated