missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Volinanserin Hydrochloride Salt, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Volinanserin Hydrochloride Salt |
| CAS | 139290-65-6 |
| Molecular Formula | C22 H28 F N O3 |
| Quantity | 1 mg |
| Synonym | MDL 100907 |
| InChI Formula | InChI=1S/C22H28FNO3/c1-26-20-5-3-4-19(22(20)27-2)21(25)17-11-14-24(15-12-17)13-10-16-6-8-18(23)9-7-16/h3-9,17,21,25H,10-15H2,1-2H3/t21-/m1/s1 |
| SMILES | COc1cccc([C@H](O)C2CCN(CCc3ccc(F)cc3)CC2)c1OC |
| IUPAC Name | (R)-(2,3-dimethoxyphenyl)-[1-[2-(4-fluorophenyl)ethyl]piperidin-4-yl]methanol |
| Molecular Weight (g/mol) | 373.46 |
| Formula Weight | 373.2053 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction