Learn More
Poulten Graf™ Volac™ USP Class A Soda Lime Glass Bulb Pipette with Works Certificate
Individually marked with the time of flow to transfer. Poulten Graf™ Volac™ USP Class A Soda Lime Glass Bulb Pipettes with Works Certificate are guaranteed to give both volumes and flow times within Class A limits.
Brand:  Poulten Graf™ US371/WAC/A/5-OLD
Product Code. 10375642
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Individually numbered and meets the accuracies of USP/ISO/BS and DIN standards
  • Amber stain graduation
  • Calibrated for delivery