Learn More
Poulten & Graf GmbH Volac™ Polystop™ Clear Dropping Bottles
Product Code. 12308208
Click to view available options
Capacity (Metric):
30 mL
50 mL
100 mL
Color:
Clear
Unit Size:
Each
Pack of 10
Product Code. 12308208
Brand: Poulten & Graf GmbH L188/A/10
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from soda lime glass. Poulten Graf™ Volac™ Polystop™ Dropping Bottles are available in two color varieties, a range of capacities and include an interchangeable 3-piece plastic stopper.
  • Stopper consists of a strong ungraduated glass pipette, a durable vinyl teat and a PP-stopper
  • Available in clear or amber glass in a variety of sizes
  • Narrow neck construction
Specifications
Capacity (Metric) 30 mL
Color Clear
Material Glass
Stopper Number 16/16
Product Type Dropping Bottle
Closure Type Stopper