missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poulten Graf™ Volac™ Glass Serological Pipettes
Offering economical single-use convenience leading to greatly reduced possibility of cross-contamination. Poulten Graf™ Volac™ Glass Serological Pipettes are disposable and have clear advantages of plastic counterparts in areas of chemical and temperature resistance, accuracy, transparency and clarity of markings.
£268.00 - £387.00
Specifications
Material | Glass |
---|---|
Graduated | Yes |
Type | Serological Pipette |
Disposable | Single-use |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10377864
|
Poulten Graf™
D808/PS |
10 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12364448
|
Poulten Graf™
D807/PS |
5 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Ideal for use with radioactive and other hazardous materials
- Volumes are color-coded
- Environmentally friendly compared with plastic counterparts
- Graduated
Specifications
Glass | |
Yes | |
Serological Pipette | |
Single-use |