Learn More
Poulten Graf™ Volac™ Glass Pasteur Pipettes
Description
- Nominal rate of delivery 50 drops/mL
- Precision made on fully automatic machines to ensure total uniformity
- Constrictions are heat formed to avoid weakness or contamination of glass
Specifications
Specifications
| Certifications/Compliance | ISO7712 |
| Material | Glass |
| Includes | Cotton Plug |
| Quantity | 1000/Pk. |
| Length (Metric) | 150 mm |
| Type | Pasteur Pipet |