Learn More
Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette
Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette allow accurate measurements of a volume of solution.
Brand:  Poulten & Graf GmbH R371/NC/A/5
Code : NEW
Additional Details : Weight : 0.15750kg
 View more versions of this product
Product Code. 15589391
  • £16.90 / Pack of 5
In Stock
to see stock.
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Amber graduation
  • With 1 mark
  • Conformity approved
  • With batch certificate