missing translation for 'onlineSavingsMsg'
Learn More

Voclosporin, MedChemExpress

Product Code. 16442946
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16442946

Brand: Medchem Express HY106638

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Voclosporin (ISAtx-247) is a calcineurin (PP2B) (CN) inhibitor.

Voclosporin (ISAtx-247) is a calcineurin (PP2B) (CN) inhibitor.

Specifications

Chemical Name or Material Voclosporin
Physical Form Solid
CAS 515814-01-4
Color White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C63H111N11O12
For Use With (Application) COVID-19-immunoregulation
Synonym ISAtx-247
Solubility Information DMSO : ≥ 50 mg/mL (41.17 mM)
SMILES C=C/C=C/C[C@@H](C)[C@H]([C@]1([H])N(C([C@](N(C([C@@H](N(C([C@](N(C([C@H](NC([C@@H](NC([C@H](CC(C)C)N(C)C([C@@](C(C)C)([H])NC([C@H](CC(C)C)N(C)C(CN(C)C([C@H](CC)NC1=O)=O)=O)=O)=O)=O)C)=O)C)=O)C)([H])CC(C)C)=O)C)CC(C)C)=O)C)([H])C(C)C)=O)C)O
Molecular Weight (g/mol) 1214.62
Formula Weight 1214.62
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.